Lanthanum powder, -200 mesh, 99.9% (REO)
Lanthanum powder, -200 mesh, 99.9% (REO)
Lanthanum powder, -200 mesh, 99.9% (REO)
Lanthanum powder, -200 mesh, 99.9% (REO)
Thermo Scientific Chemicals
Lanthanum powder, -200 mesh, 99.9% (REO)
CAS: 7439-91-0 | La | 138.91 g/mol
Have Questions?
Catalog NumberQuantity
ALF045908.065 g
Catalog number ALF045908.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialLanthanum powder
ColorSilver
CAS7439-91-0
Health Hazard 1H228
Health Hazard 2GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under Argon
RUO – Research Use Only