Silicone oil, high temperature, usable temperature range: 25 to 250°C (open system) and 25 to 315°C (closed system)
Silicone oil, high temperature, usable temperature range: 25 to 250°C (open system) and 25 to 315°C (closed system)
Silicone oil, high temperature, usable temperature range: 25 to 250°C (open system) and 25 to 315°C (closed system)
Silicone oil, high temperature, usable temperature range: 25 to 250°C (open system) and 25 to 315°C (closed system)
Thermo Scientific Chemicals
Silicone oil, high temperature, usable temperature range: 25 to 250°C (open system) and 25 to 315°C (closed system)
CAS: 68083-14-7 | C16H22O2Si2 | 302.52 g/mol
Have Questions?
Catalog NumberQuantity
ALF045896.36500 g
Catalog number ALF045896.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSilicone Oil
Name NoteHigh temperature, Usable temp. range: 25 to 250°C (open system) and 25 to 315°C (closed system)
CAS68083-14-7
ColorColorless
Recommended StorageAmbient temperatures
View more
Silicone oil is used in high temperature open systems, closed systems and circulating, closed loop and heat transfer baths. It is used as a high temperature fluid for laboratory research apparatus and instruments. It is also used as a calibration media in temperature controlled baths.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • High temperature open system, closed system, circulating, closed loop, and heat transfer baths
  • High temperature fluids for laboratory research apparatus and instruments
RUO – Research Use Only