Zeolite ferrierite, ammonium
Zeolite ferrierite, ammonium
Zeolite ferrierite, ammonium
Zeolite ferrierite, ammonium
Thermo Scientific Chemicals
Zeolite ferrierite, ammonium
CAS: 1318-02-1 | Al2O5Si | 162.043 g/mol
Have Questions?
Catalog NumberQuantity
ALF045884.22100 g
Catalog number ALF045884.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialZeolite ferrierite, ammonium
CAS1318-02-1
ColorWhite
Recommended StorageAmbient temperatures
Physical FormPowder
View more
Zeolite ferrierite, ammonium is used as a commercial filter and ion-exchange bed. Further, it acts as a catalyst for the acid-catalyzed isomerization of n-butenes to isobutene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Zeolite ferrierite is used as commercial filters, ion-exchange beds, and as a catalyst for the acid-catalyzed isomerization of n-butenes to isobutene
RUO – Research Use Only