Zeolite mordenite, ammonium
Zeolite mordenite, ammonium
Zeolite mordenite, ammonium
Zeolite mordenite, ammonium
Thermo Scientific Chemicals
Zeolite mordenite, ammonium
Zeolite mordenite is used for the acid-catalyzed isomerization of alkanes and aromatics | CAS: 1318-02-1 | Al2O5Si | 162.043 g/mol
Have Questions?
Catalog NumberQuantity
ALF045877.22100 g
Catalog number ALF045877.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialZeolite mordenite, ammonium
CAS1318-02-1
ColorWhite
Recommended StorageAmbient temperatures
Physical FormPowder
View more
Zeolite mordenite is used for the acid-catalyzed isomerization of alkanes and aromatics. Used in detergent manufacturing for the applications of the removal of hardness. It is also used as Odor control, industrial absorbents, industrial fillers, gas absorption, soil remediation, wastewater filtration, flocculating agent, animal feeds, hydroponics, molecular sieves, desiccants, lightweight concrete, acoustics and ceramics.