Zeolite beta, ammonium
Zeolite beta, ammonium
Zeolite beta, ammonium
Zeolite beta, ammonium
Thermo Scientific Chemicals
Zeolite beta, ammonium
CAS: 1318-02-1 | Al2O5Si | 162.043 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number ALF045873.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialZeolite beta, ammonium
CAS1318-02-1
ColorWhite
Recommended StorageAmbient temperatures
Physical FormPowder
View more
Zeolite beta is used in isomerization of waxes, and Friedel Crafts reactions. Used in detergent manufacturing for the applications of the removal of hardness. It is also used as Odor control, industrial absorbents, industrial fillers, gas absorption, soil remediation, wastewater filtration, flocculating agent, animal feeds, hydroponics, molecular sieves, desiccants, lightweight concrete, acoustics and ceramics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Zeolite beta is used in isomerization of waxes, and Friedel Crafts reactions (alkylation and acylation)
RUO – Research Use Only