Potassium, solid, 99% (metals basis)
Potassium, solid, 99% (metals basis)
Potassium, solid, 99% (metals basis)
Potassium, solid, 99% (metals basis)
Thermo Scientific Chemicals
Potassium, solid, 99% (metals basis)
CAS: 7440-09-7 | K | 39.10 g/mol
Have Questions?
Catalog NumberQuantity
ALF045842.065 g
Catalog number ALF045842.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium
CAS7440-09-7
Health Hazard 1H260-H314-H335
Health Hazard 2GHS H Statement
H260-H314-H318
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P223-P231+P232-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
View more
Potassium combines under pressure with silver metal to form 2 new compounds: K2Ag and K3Ag. This chemical is important in neuron (brain and nerve) function, and in influencing osmotic balance between cells and the interstitial fluid, with their distribution mediated in all animals (but not in all plants) by the so-called Na+/K+-ATPase pump. It is also used to prevent muscle contraction and the sending of all nerve impulses in animals through action potentials. Potassium ion is a nutrient necessary for human life and health. It is used as reducing agent in organic synthesis.