Gallium metal, packaged in polyethylene bottle, 99.9% (metals basis)
Gallium metal, packaged in polyethylene bottle, 99.9% (metals basis)
Gallium metal, packaged in polyethylene bottle, 99.9% (metals basis)
Gallium metal, packaged in polyethylene bottle, 99.9% (metals basis)
Thermo Scientific Chemicals
Gallium metal, packaged in polyethylene bottle, 99.9% (metals basis)
May be solid or liquid due to ambient temperature | CAS: 7440-55-3 | Ga | 69.72 g/mol
Have Questions?
Catalog NumberQuantity
ALF045841.36500 g
Catalog number ALF045841.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialGallium
ColorSilver to Gray
CAS7440-55-3
Health Hazard 1H290-H314-H335
Health Hazard 3P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c