Silver Conductive Ink
Silver Conductive Ink
Thermo Scientific Chemicals
Silver Conductive Ink
Have Questions?
Catalog NumberQuantity
ALF045661.065 g
Catalog number ALF045661.06
Price (MYR)
278.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
278.00
EA
Add to cart
Specifications
Chemical Name or MaterialSilver Conductive Ink
Health Hazard 1H319
Health Hazard 3P264b-P280i-P305+P351+P338
Density2.5