Devarda's Alloy, granular
Devarda's Alloy, granular
Devarda's Alloy, granular
Devarda's Alloy, granular
Thermo Scientific Chemicals
Devarda's Alloy, granular
CAS: 8049-11-4 | Al9Cu10Zn | 943.67 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF045526.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDevarda's Alloy
CAS8049-11-4
Health Hazard 1H228-H500
Health Hazard 3P210-P240-P241-P280-P370+P378q
Recommended StorageAmbient temperatures
View more
Devarda's Alloy is used as reducing agent in analytical chemistry for the determination of nitrates after their reduction to ammonia under alkaline conditions. It is also used in the quantitative or qualitative analysis of nitrates in agriculture and soil science before the development of ion chromatography. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, and agrochemicals.