Silver nanopowder, APS 20-40nm, 99.9% (metals basis)
Silver nanopowder, APS 20-40nm, 99.9% (metals basis)
Silver nanopowder, APS 20-40nm, 99.9% (metals basis)
Silver nanopowder, APS 20-40nm, 99.9% (metals basis)
Thermo Scientific Chemicals
Silver nanopowder, APS 20-40nm, 99.9% (metals basis)
CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALF045509.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSilver nanopowder
Name NoteNo Oxide layer
CAS7440-22-4
Recommended StorageAmbient temperatures
DOT InformationTransport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
View more
Silver nanopowder is involved in the coating of silver and is used for medicinal purposes and in medical devices. Its larger surface area to volume ratio and greater activity of nano-silver are utilized in modern biochemical applications and drug delivery. It is also used in the fabrication of graphite epoxy nanocomposites and optical fiber sensors. Furthermore, it is involved in the formulation of water based ink.