Manganese nanopowder, APS 30-50nm
Manganese nanopowder, APS 30-50nm
Manganese nanopowder, APS 30-50nm
Manganese nanopowder, APS 30-50nm
Thermo Scientific Chemicals
Manganese nanopowder, APS 30-50nm
Form: Mn3O4 layer 5-12nm | CAS: 7439-96-5 | Mn | 54.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALF045508.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialManganese
ColorGray
CAS7439-96-5
Health Hazard 1H228-H319
Health Hazard 2Take precautionary measures against static discharges.