Titanium wire, 1.0mm (0.04in) dia, hard, 99.99% (metals basis)
Titanium wire, 1.0mm (0.04in) dia, hard, 99.99% (metals basis)
Titanium wire, 1.0mm (0.04in) dia, hard, 99.99% (metals basis)
Titanium wire, 1.0mm (0.04in) dia, hard, 99.99% (metals basis)
Thermo Scientific Chemicals
Titanium wire, 1.0mm (0.04in) dia, hard, 99.99% (metals basis)
CAS: 7440-32-6 | Ti | 47.87 g/mol
Have Questions?
Catalog NumberQuantity
ALF045485.BY100 cm
Catalog number ALF045485.BY
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 cm
Request bulk or custom format
Specifications
Chemical Name or MaterialTitanium wire
Name Note1.0mm (0.04 in.) dia., Hard
CAS7440-32-6
Recommended StorageAmbient temperatures
EINECS Number231-142-3