Carbon, activated, -20+40 mesh
Carbon, activated, -20+40 mesh
Carbon, activated, -20+40 mesh
Carbon, activated, -20+40 mesh
Thermo Scientific Chemicals
Carbon, activated, -20+40 mesh
CAS: 7440-44-0 | C | 12.011 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
100 g
500 g
Catalog number ALF045478.A3
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialCarbon
CAS7440-44-0
Health Hazard 1H500
ColorBlack