Aluminum foil, alloy 6061, 2.0mm (0.08in) thick
Aluminum foil, alloy 6061, 2.0mm (0.08in) thick
Thermo Scientific Chemicals
Aluminum foil, alloy 6061, 2.0mm (0.08in) thick
Have Questions?
Catalog NumberQuantity
ALF045171.HB300 x 300 mm
Catalog number ALF045171.HB
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
300 x 300 mm
Request bulk or custom format
Specifications
Chemical Name or MaterialAluminum foil, Alloy 6061
Name Note2.0mm (0.08 in.) thick
Recommended StorageAmbient temperatures
TSCANo
MDL NumberMFCD00134029
View more
Aluminum foil, alloy 6061 is commonly used for structural components, frames, brackets, jigs, fixtures, base plates, machine parts, hydraulic valve bodies, valves parts, fuse parts, gears, worm gears, rectifier parts, fasteners, truck and marine components, marine fittings and hardware, electrical fittings and connectors, hinge pins, magneto parts, appliance fittings, camera lens and mounts 6061 is used for heavy duty structures requiring good strength-to-weight ratio with good corrosion resistance. 6061 is easily cold worked and formed in the annealed condition. Cutting, stamping, bending, deep drawing, drilling and tapping. are all readily accomplished using standard methods.