Sulfuric acid, 0.2N Standardized Solution
Sulfuric acid, 0.2N Standardized Solution
Sulfuric acid, 0.2N Standardized Solution
Sulfuric acid, 0.2N Standardized Solution
Sulfuric acid, 0.2N Standardized Solution
Thermo Scientific Chemicals
Sulfuric acid, 0.2N Standardized Solution
CAS: 7664-93-9 | H2O4S | 98.07 g/mol
Have Questions?
Catalog NumberQuantity
ALF045062.K21 L
Catalog number ALF045062.K2
Price (MYR)
373.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
373.00
EA
Add to cart
Specifications
Chemical Name or MaterialSulfuric acid, 0.2N Standardized Solution
CAS7664-93-9
Recommended StorageAmbient temperatures
RTECS NumberWS5600000
EINECS Number231-639-5
View more
Sulfuric acid, 0.2N standardized solution is used for volumetric analysis in laboratories.