Sulfuric acid, 0.05N Standardized Solution
Sulfuric acid, 0.05N Standardized Solution
Sulfuric acid, 0.05N Standardized Solution
Sulfuric acid, 0.05N Standardized Solution
Thermo Scientific Chemicals
Sulfuric acid, 0.05N Standardized Solution
CAS: 7664-93-9 | H2O4S | 98.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
1 L
Catalog number ALF045048.K7
Price (MYR)
479.00
EA
Add to cart
Quantity:
4 L
Request bulk or custom format
Price (MYR)
479.00
EA
Add to cart
Specifications
AppearanceColourless solution
Chemical Name or MaterialSulfuric acid
ColorColorless
CAS7664-93-9
CAS7732-18-5