Tin ingot, 99.998% (metals basis)
Tin ingot, 99.998% (metals basis)
Tin ingot, 99.998% (metals basis)
Tin ingot, 99.998% (metals basis)
Tin ingot, 99.998% (metals basis)
CAS: 7440-31-5 | Sn | 118.71 g/mol
Have Questions?
Catalog NumberQuantity
ALF045032.A11 kg
Catalog number ALF045032.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialTin
CAS7440-31-5
AppearanceSilver Gray ingot
Recommended StorageAmbient temperatures
EINECS Number231-141-8