Indium powder, -100 mesh, 99.99% (metals basis)
Indium powder, -100 mesh, 99.99% (metals basis)
Indium powder, -100 mesh, 99.99% (metals basis)
Indium powder, -100 mesh, 99.99% (metals basis)
Thermo Scientific Chemicals
Indium powder, -100 mesh, 99.99% (metals basis)
CAS: 7440-74-6 | In | 114.82 g/mol
Have Questions?
Catalog NumberQuantity
ALF044986.22100 g
Catalog number ALF044986.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialIndium powder
ColorSilver to Gray
CAS7440-74-6
Melting Point156.6°C
Boiling Point2072°C