Cerium(IV) oxide, nanopowder, 99.5% min (REO)
Cerium(IV) oxide, nanopowder, 99.5% min (REO)
Cerium(IV) oxide, nanopowder, 99.5% min (REO)
Cerium(IV) oxide, nanopowder, 99.5% min (REO)
Thermo Scientific Chemicals
Cerium(IV) oxide, nanopowder, 99.5% min (REO)
CAS: 1306-38-3 | CeO2 | 172.114 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALF044960.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCerium(IV) oxide
Melting Point2600°C
Name NoteRare Earth Oxides Content
CAS1306-38-3
ColorYellow
View more
Cerium(IV) oxide is widely applied as a solid electrolyte in fuel cells. It is also used in ceramics and to polish glass and stones. It can also be employed as a co-catalyst in many reactions. It is also used as an antioxidant and to generate hydrogen through water splitting. It is used as a high-temperature hydrocarbon oxidation catalyst in self-cleaning ovens. It can be used in infrared filters, incandescent mantles and sunscreens in combination with other materials. It finds use as a catalyst support and to sensitize photosensitive glass. It is utilized to increase specific surface area and inhibit sintering as well as pore size growth at a high temperature by addition with CuO/TiO2.