Tetramethylammonium hydroxide, 1.0 M aq. soln., ACS
Tetramethylammonium hydroxide, 1.0 M aq. soln., ACS
Tetramethylammonium hydroxide, 1.0 M aq. soln., ACS
Tetramethylammonium hydroxide, 1.0 M aq. soln., ACS
Thermo Scientific Chemicals
Tetramethylammonium hydroxide, 1.0 M aq. soln., ACS
CAS: 75-59-2 | C4H13NO | 91.154 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Catalog number ALF044938.K2
Price (MYR)
1,206.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
1,206.00
EA
Add to cart
Specifications
Chemical Name or MaterialTetramethylammonium hydroxide
CAS75-59-2
Health Hazard 1H302-H311-H314-H370-H372
Health Hazard 2GHS H Statement
H301-H311-H314-H371-H373
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
May cause damage to organs.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
View more
Tetramethylammonium hydroxide is used as anisotropic etching of silicon, as a basic solvent in the development of acidic photo resist in the photolithography process, and is highly effective in stripping photo resist, and is used as a surfactant in the synthesis of ferrofluid, to inhibit nanoparticle aggregation.