Nickel foil, 2.0mm (0.08in) thick, 99.5% (metals basis)
Nickel foil, 2.0mm (0.08in) thick, 99.5% (metals basis)
Nickel foil, 2.0mm (0.08in) thick, 99.5% (metals basis)
Nickel foil, 2.0mm (0.08in) thick, 99.5% (metals basis)
Thermo Scientific Chemicals
Nickel foil, 2.0mm (0.08in) thick, 99.5% (metals basis)
CAS: 7440-02-0 | Ni | 58.69 g/mol
Have Questions?
Catalog NumberQuantity
ALF044825.GH100 x 100 mm
Catalog number ALF044825.GH
Price (MYR)
948.00
EA
Add to cart
Quantity:
100 x 100 mm
Request bulk or custom format
Price (MYR)
948.00
EA
Add to cart
Specifications
Chemical Name or MaterialNickel foil
Name Note2.0mm (0.08 in.) thick
CAS7440-02-0
Health Hazard 1H317-H351-H372
Health Hazard 2GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.