Nickel foil, 1.0mm (0.04in) thick, 99.5% (metals basis)
Nickel foil, 1.0mm (0.04in) thick, 99.5% (metals basis)
Nickel foil, 1.0mm (0.04in) thick, 99.5% (metals basis)
Nickel foil, 1.0mm (0.04in) thick, 99.5% (metals basis)
Nickel foil, 1.0mm (0.04in) thick, 99.5% (metals basis)
Thermo Scientific Chemicals
Nickel foil, 1.0mm (0.04in) thick, 99.5% (metals basis)
Form: Approx.22.3g/50 x 50mm | CAS: 7440-02-0 | Ni | 58.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 x 100 mm
50 x 50 mm
Catalog number ALF044824.GH
Price (MYR)
679.00
EA
Add to cart
Quantity:
100 x 100 mm
Request bulk or custom format
Price (MYR)
679.00
EA
Add to cart
Specifications
Chemical Name or MaterialNickel
ColorSilver to Gray
CAS7440-02-0
Health Hazard 1H317-H351-H372
AppearanceSilver gray foil