Cyclopentane, HPLC Grade
Cyclopentane, HPLC Grade
Cyclopentane, HPLC Grade
Cyclopentane, HPLC Grade
Thermo Scientific Chemicals
Cyclopentane, HPLC Grade
For HPLC and Spectrophotometry | CAS: 287-92-3 | C5H10 | 70.135 g/mol
Have Questions?
Catalog NumberQuantity
ALF044814.K21 L
Catalog number ALF044814.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialCyclopentane
Name NoteContains 75+% cyclopentane. Remainder consists of C5 and C6 hydrocarbons .
CAS287-92-3
Health Hazard 1H225-H304-H335-H336
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P301+P310-P303+P361+P353-P304+P340-P312-P331-P370+P378q-P501c
View more
Cyclopentane is used in all analytical laboratories as a major component of mobile phase in High Performance Liquid Chromatography (HPLC) and in Liquid Chromatography - Mass Spectrometry (LC-MS).