12-Hydroxystearic acid, 95%
12-Hydroxystearic acid, 95%
12-Hydroxystearic acid, 95%
12-Hydroxystearic acid, 95%
Thermo Scientific Chemicals
12-Hydroxystearic acid, 95%
CAS: 106-14-9 | C18H36O3 | 300.483 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number ALF044810.14
Price (MYR)
2,022.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
2,022.00
EA
Add to cart
Specifications
Chemical Name or Material12-Hydroxystearic acid
CAS106-14-9
Melting Point72°C to 76°C
ColorWhite
Recommended StorageAmbient temperatures
View more
12-Hydroxystearic acid is used in cosmetics, wax blends, heavy duty greases, polishes, inks and hot melt adhesives, as a lubricant for natural and synthetic rubbers and as a source for industrial oleo chemicals. It is also used as an intermediate in pharmaceuticals.