Silicon(IV) oxide, amorphous, nanopowder, 99.9% (metals basis)
Silicon(IV) oxide, amorphous, nanopowder, 99.9% (metals basis)
Silicon(IV) oxide, amorphous, nanopowder, 99.9% (metals basis)
Silicon(IV) oxide, amorphous, nanopowder, 99.9% (metals basis)
Thermo Scientific Chemicals
Silicon(IV) oxide, amorphous, nanopowder, 99.9% (metals basis)
CAS: 7631-86-9 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number ALF044781.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Boiling Point2230°C
Chemical Name or MaterialSilicon(IV) oxide
Melting Point1710°C
CAS7631-86-9
ColorWhite
View more
Silicon(IV) oxide is used as sodium aluminate stabilizer. It is also used as SDS mixture of sodium alkyl sulfates consisting chiefly of sodium lauryl sulfate. It is also acts as chain transfer agent in polymerization.