Erbium(III) oxide, nanopowder, 99.5% (REO)
Erbium(III) oxide, nanopowder, 99.5% (REO)
Erbium(III) oxide, nanopowder, 99.5% (REO)
Erbium(III) oxide, nanopowder, 99.5% (REO)
Thermo Scientific Chemicals
Erbium(III) oxide, nanopowder, 99.5% (REO)
S. A. 13 to 17m2/g | CAS: 12061-16-4 | Er2O3 | 382.52 g/mol
Have Questions?
Catalog NumberQuantity
ALF044668.1850 g
Catalog number ALF044668.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialErbium(III) oxide
Melting Point2400°C
CAS12061-16-4
Health Hazard 1H319: Causes serious eye irritation.