Silicon carbide, beta-phase, nanopowder
Silicon carbide, beta-phase, nanopowder
Silicon carbide, beta-phase, nanopowder
Silicon carbide, beta-phase, nanopowder
Thermo Scientific Chemicals
Silicon carbide, beta-phase, nanopowder
S.A. 70 to 90 m2/g | CAS: 409-21-2 | CSi | 40.10 g/mol
Have Questions?
Catalog NumberQuantity
ALF044646.1425 g
Catalog number ALF044646.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSilicon carbide
Melting Point2700°C
Name Notebeta-phase
CAS409-21-2
ColorBlack to Gray
View more
Silicon carbide can be used form very hard ceramics that are widely used in applications requiring high endurance, such as car brakes, car clutches and ceramic plates in bulletproof vests. Electronic applications of silicon carbide as light-emitting diodes (LEDs) and detectors in early radios.