Tellurium, plasma standard solution, Specpure™ Te 1000μg/mL
Tellurium, plasma standard solution, Specpure™ Te 1000μg/mL
Thermo Scientific Chemicals
Tellurium, plasma standard solution, Specpure™ Te 1000μg/mL
Te in 5% HNO3
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
50 mL
Catalog number ALF044632.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialTellurium, plasma standard solution, Specpure™
Health Hazard 1H290-H314-H335
Health Hazard 2GHS H Statement
H314-H318;
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c