Ethylene glycol, Spectrophotometric grade, 99+%
Ethylene glycol, Spectrophotometric grade, 99+%
Ethylene glycol, Spectrophotometric grade, 99+%
Ethylene glycol, Spectrophotometric grade, 99+%
Thermo Scientific Chemicals
Ethylene glycol, Spectrophotometric grade, 99+%
CAS: 107-21-1 | C2H6O2 | 62.068 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 x 1 L
1 L
Catalog number ALF044529.M1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
4 x 1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialEthylene glycol
CAS107-21-1
Health Hazard 1H302-H336-H373
Health Hazard 3P260-P264b-P270-P271-P301+P312-P304+P340-P312-P314-P330-P501c
Melting Point°C to 13°C
View more
For use in spectrophotometryEthylene glycol is used in spectrophotometry and environmental testing.