Nitric acid, 2.0N Standardized Solution
Nitric acid, 2.0N Standardized Solution
Nitric acid, 2.0N Standardized Solution
Nitric acid, 2.0N Standardized Solution
Thermo Scientific Chemicals
Nitric acid, 2.0N Standardized Solution
CAS: 7697-37-2 | HNO3 | 63.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
Catalog number ALF044528.K2
Price (MYR)
2,200.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
2,200.00
EA
Add to cart
Specifications
Chemical Name or MaterialNitric Acid, 2N Standardized Solution
CAS7697-37-2
Health Hazard 1H290-H314-H332
Density1.06 g/mL
Health Hazard 3P234-P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
View more
Nitric acid, 2.0N Standardized Solution is used for volumetric analysis in laboratories.