Amidosulfonic acid, 99.99% (metals basis)
Amidosulfonic acid, 99.99% (metals basis)
Amidosulfonic acid, 99.99% (metals basis)
Amidosulfonic acid, 99.99% (metals basis)
Thermo Scientific Chemicals
Amidosulfonic acid, 99.99% (metals basis)
CAS: 5329-14-6 | H3NO3S | 97.088 g/mol
Have Questions?
Catalog NumberQuantity
ALF044510.1425 g
Catalog number ALF044510.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmidosulfonic acid
pH1.2
CAS5329-14-6
Melting Point∼205°C (decomposition)
Health Hazard 1H290-H315-H319
View more
Amidosulfonic acid is mainly a precursor to sweet-tasting compounds. Reaction with cyclohexylamine followed by addition of NaOH gives C6H11NHSO3Na, sodium cyclamate. Sulfamic acid is used as an acidic cleaning agent