Perchloric acid, ACS, 48-50%
Perchloric acid, ACS, 48-50%
Perchloric acid, ACS, 48-50%
Perchloric acid, ACS, 48-50%
Thermo Scientific Chemicals
Perchloric acid, ACS, 48-50%
Maximum level of impurities: Color (APHA) 10, Residue after ignition 0.003%, Silicate and phosphate as SiO2 5ppm, Cl 0.001%, Nitrogen compounds (as N) 0.001%, SO4 0.001%, Heavy Metals (as Pb) 1ppm, Fe 1ppm | CAS: 7601-90-3 | ClHO4 | 100.45 g/mol
Have Questions?
Catalog NumberQuantity
ALF044464.AP500 mL
Catalog number ALF044464.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialPerchloric acid
CAS7601-90-3
Melting Point°C to 112°C
Health Hazard 1H271-H290-H302-H314-H335-H373
Density1.41 g/cm3 at 20°C
View more
Perchloric acid is used as a precursor for ammonium perchlorate, which is used in rocket fuel. It is useful for etching of liquid crystal displays, etching of chrome, ore extraction and in analytical chemistry. It is used as a strong oxidizer and as a solvent for metals and alloys. It serves as a deproteinization reagent in food analysis. As a dehydrating agent, it is used for the determination of silica in iron and steel and in cement and other silicate materials. It is involved in the separation and determination of copper and other metals and alkali metals such as sodium and potassium. It is also used as an adjunct to increase the reduction potential of cerate salts in volumetric analysis.