Cerium(III) acetate hydrate, 99.995% (metals basis)
Cerium(III) acetate hydrate, 99.995% (metals basis)
Cerium(III) acetate hydrate, 99.995% (metals basis)
Cerium(III) acetate hydrate, 99.995% (metals basis)
Cerium(III) acetate hydrate, 99.995% (metals basis)
Thermo Scientific Chemicals
Cerium(III) acetate hydrate, 99.995% (metals basis)
CAS: 206996-60-3 | C6H9CeO6 | 317.25 g/mol
Have Questions?
Catalog NumberQuantity
ALF044433.0910 g
Catalog number ALF044433.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCerium (III) acetate hydrate
CAS206996-60-3
ColorWhite
Health Hazard 1H315-H318-H335
Health Hazard 2GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
View more
Cerium(III) acetate hydrate is used in thin fim buffer layers suitable for high κ dielectrics by sol-gel route. And it is also used as a catalyst for auto-oxidation of cresols.