Copper(II) acetate, 99.999% (metals basis)
Copper(II) acetate, 99.999% (metals basis)
Copper(II) acetate, 99.999% (metals basis)
Copper(II) acetate, 99.999% (metals basis)
Thermo Scientific Chemicals
Copper(II) acetate, 99.999% (metals basis)
CAS: 142-71-2 | C4H6CuO4 | 181.634 g/mol
Have Questions?
Catalog NumberQuantity
ALF044355.22100 g
Catalog number ALF044355.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCopper(II) acetate
CAS142-71-2
ColorGreen
Health Hazard 1H302-H314-H335-H373
Health Hazard 2In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.
View more
Copper(II) acetate is used in biochemical applications such as DNA extraction. It is used as a source of copper in inorganic synthesis, an oxidizing agent and catalyst in organic synthesis. It is also used to couple two terminal alkynes to make a 1,3-diyne. It is widely used in pigments, manufacture of paris green, textile dyeing, skin conditioner in cosmetics, antioxidant and stabilizer in food grade polymers.