Gallium pellets, 6mm dia, 99.99999% (metals basis)
Gallium pellets, 6mm dia, 99.99999% (metals basis)
Gallium pellets, 6mm dia, 99.99999% (metals basis)
Gallium pellets, 6mm dia, 99.99999% (metals basis)
Thermo Scientific Chemicals
Gallium pellets, 6mm dia, 99.99999% (metals basis)
CAS: 7440-55-3 | Ga | 69.72 g/mol
Have Questions?
Catalog NumberQuantity
ALF044325.0910 g
Catalog number ALF044325.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or MaterialGallium pellets
ColorGray-Silver
Name Note6mm dia.
CAS7440-55-3
Health Hazard 1H290-H314-H335