Gallium pellets, 99.9999% (metals basis)
Gallium pellets, 99.9999% (metals basis)
Gallium pellets, 99.9999% (metals basis)
Gallium pellets, 99.9999% (metals basis)
Thermo Scientific Chemicals
Gallium pellets, 99.9999% (metals basis)
CAS: 7440-55-3 | Ga | 69.72 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number ALF044324.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialGallium
CAS7440-55-3
Health Hazard 1H290-H314-H335
Health Hazard 3P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
Recommended StorageKeep cold