Silicon powder, APS 1-5 micron, 99.9% (metals basis)
Silicon powder, APS 1-5 micron, 99.9% (metals basis)
Silicon powder, APS 1-5 micron, 99.9% (metals basis)
Silicon powder, APS 1-5 micron, 99.9% (metals basis)
Silicon powder, APS 1-5 micron, 99.9% (metals basis)
Thermo Scientific Chemicals
Silicon powder, APS 1-5 micron, 99.9% (metals basis)
CAS: 7440-21-3 | Si | 28.09 g/mol
Have Questions?
Catalog NumberQuantity
ALF044185.0910 g
Catalog number ALF044185.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSilicon
Name NoteFor analysis APS
CAS7440-21-3
Health Hazard 1H228
Health Hazard 3P210-P240-P241-P280-P370+P378q