1-Methyl-2-pyrrolidinone, Biograde, 99.5%
1-Methyl-2-pyrrolidinone, Biograde, 99.5%
1-Methyl-2-pyrrolidinone, Biograde, 99.5%
1-Methyl-2-pyrrolidinone, Biograde, 99.5%
1-Methyl-2-pyrrolidinone, Biograde, 99.5%
Thermo Scientific Chemicals
1-Methyl-2-pyrrolidinone, Biograde, 99.5%
Suitable for Biotech applications. Useful for spectrophotometry, chromatography and ICP-MS detection. Also, used for the chemical functionalization of graphene sheets by solvothermal reduction of graphene oxide. | CAS: 872-50-4 | C5H9NO | 99.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
ALF044063.K21 LUnder Argon
Catalog number ALF044063.K2
Price (MYR)
800.00
EA
Add to cart
Quantity:
1 L
Packaging:
Under Argon
Request bulk or custom format
Price (MYR)
800.00
EA
Add to cart
Specifications
Chemical Name or Material1-Methyl-2-pyrrolidinone
Melting Point-24°C
CAS872-50-4
ColorColorless
Health Hazard 1H227-H315-H319-H335-H360D-H373
View more
Suitable for Biotech applications.1-methyl-2-pyrrolidinone, is useful for spectrophotometry, chromatography and ICP-MS detection. It is useful for the chemical functionalization of graphene sheets by solvothermal reduction of graphene oxide. It is also useful for pharmaceutical and different industrial applications due to its broad solvency and low volatility.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Suitable for Biotech applications
  • Useful for spectrophotometry, chromatography and ICP-MS detection
  • Used for the chemical functionalization of graphene sheets by solvothermal reduction of graphene oxide
  • Also useful for pharmaceutical, different industrial applications due to its broad solvency and low volatility
RUO – Research Use Only