Copper turnings, 99.9% (metals basis)
Copper turnings, 99.9% (metals basis)
Copper turnings, 99.9% (metals basis)
Copper turnings, 99.9% (metals basis)
Thermo Scientific Chemicals
Copper turnings, 99.9% (metals basis)
Typically 1-12mm wide | CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Catalog NumberQuantity
ALF044001.1850 g
Catalog number ALF044001.18
Price (MYR)
390.00
EA
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
390.00
EA
Add to cart
Specifications
Chemical Name or MaterialCopper
CAS7440-50-8
AppearanceCopper colored turnings
Recommended StorageAmbient temperatures
Quantity50 g