Cerium ingot, 99% (REO)
Cerium ingot, 99% (REO)
Cerium ingot, 99% (REO)
Cerium ingot, 99% (REO)
Thermo Scientific Chemicals
Cerium ingot, 99% (REO)
CAS: 7440-45-1 | Ce | 140.12 g/mol
Have Questions?
Catalog NumberQuantity
ALF043977.30250 g
Catalog number ALF043977.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCerium ingot
ColorGray
CAS7440-45-1
Health Hazard 1H228
Health Hazard 2GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under mineral oil
RUO – Research Use Only