Cyanide, standard solution, Specpure™ CN∣- 1000μg/mL
Cyanide, standard solution, Specpure™ CN∣- 1000μg/mL
Thermo Scientific Chemicals
Cyanide, standard solution, Specpure™ CN∣- 1000μg/mL
KCN in 0.1% NaOH
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALF043804.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialCyanide
Name NoteStandard Solution
Health Hazard 1H301-H312
Health Hazard 2Keep container in a well ventilated place. Wear suitable protective clothing and gloves.
Health Hazard 3P264b-P270-P280-P301+P310-P302+P352-P312-P330-P363-P501c
View more
Cyanide, standard solution is used to dissolve metals and their ores. It is also used in clinical chemistry and in waste water treatment facilities to determine the concentration of dissolved and particulate materials.