1,6-Diisocyanatohexane, 98+%
1,6-Diisocyanatohexane, 98+%
1,6-Diisocyanatohexane, 98+%
1,6-Diisocyanatohexane, 98+%
Thermo Scientific Chemicals
1,6-Diisocyanatohexane, 98+%
CAS: 822-06-0 | C8H12N2O2 | 168.20 g/mol
Have Questions?
Catalog NumberQuantity
ALF043803.30250 g
Catalog number ALF043803.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or Material1,6-Diisocyanatohexane
CAS822-06-0
Health Hazard 1H302-H314-H317-H330-H334-H335
Health Hazard 2GHS H Statement
H331-H334-H315-H319-H317-H335-H204
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Fire or projection hazard.
Health Hazard 3P260-P264b-P270-P271-P272-P280-P284-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
View more
1,6-Diisocyanatohexane is used as a polymerizing agent in polyurethane paints and coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under Argon
RUO – Research Use Only