Nickel powder, spherical, -400 mesh, 99.5+% (metals basis), Thermo Scientific Chemicals
Nickel powder, spherical, -400 mesh, 99.5+% (metals basis), Thermo Scientific Chemicals
Nickel powder, spherical, -400 mesh, 99.5+% (metals basis), Thermo Scientific Chemicals
Nickel powder, spherical, -400 mesh, 99.5+% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Nickel powder, spherical, -400 mesh, 99.5+% (metals basis), Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantity
ALF043704.36500 g
Catalog number ALF043704.36
Price (MYR)
1,063.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
1,063.00
EA
Add to cart
Specifications
Chemical Name or MaterialNickel powder
CAS7440-02-0
Health Hazard 1H228-H317-H350-H372
Health Hazard 2GHS H Statement
H372-H351-H228-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
Flammable solid.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P210-P240-P241-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P370+P378q-P501c