Copper(II) phthalocyanine
Copper(II) phthalocyanine
Copper(II) phthalocyanine
Copper(II) phthalocyanine
Thermo Scientific Chemicals
Copper(II) phthalocyanine
CAS: 147-14-8 | C32H16CuN8
Have Questions?
Catalog NumberQuantity
ALF043650.1850 g
Catalog number ALF043650.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCopper (II) phthalocyanine
CAS147-14-8
Melting Point350°C
ColorBlue to Purple
Recommended StorageAmbient temperatures
View more
Copper(II) phthalocyanine are involved in the study of photosensitizer chemistry for uniform polymerization, luminescence chemistry and spectrophotometric analysis, organic synthesis and polymerization. This pigment is used in enamels, linoleum, inks, plastics, and rubber goods. Photoisomerizable phthalocyanines are used in rewritable CD or DVD printing. Other applications in organic solar cells, biosensitizers and display devices such as OLED , OTFT, Wearable Display, and e-paper.