Titanium plate, 3.175mm (0.125in) thick, 100x100mm (3.9x3.9in), 99.99% (metals basis)
Titanium plate, 3.175mm (0.125in) thick, 100x100mm (3.9x3.9in), 99.99% (metals basis)
Titanium plate, 3.175mm (0.125in) thick, 100x100mm (3.9x3.9in), 99.99% (metals basis)
Titanium plate, 3.175mm (0.125in) thick, 100x100mm (3.9x3.9in), 99.99% (metals basis)
Thermo Scientific Chemicals
Titanium plate, 3.175mm (0.125in) thick, 100x100mm (3.9x3.9in), 99.99% (metals basis)
CAS: 7440-32-6 | Ti | 47.87 g/mol
Have Questions?
Catalog NumberQuantity
ALF043361.KG1 Pc.
Catalog number ALF043361.KG
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 Pc.
Request bulk or custom format
Specifications
Chemical Name or MaterialTitanium plate
Name Note3.175mm (0.125 in.) thick, 100x100mm (3.9x3.9 in.)
CAS7440-32-6
Recommended StorageAmbient temperatures
EINECS Number231-142-3