1,6-Hexanediol diacrylate, 99% (reactive esters), stab. with 90ppm hydroquinone
1,6-Hexanediol diacrylate, 99% (reactive esters), stab. with 90ppm hydroquinone
1,6-Hexanediol diacrylate, 99% (reactive esters), stab. with 90ppm hydroquinone
1,6-Hexanediol diacrylate, 99% (reactive esters), stab. with 90ppm hydroquinone
Thermo Scientific Chemicals
1,6-Hexanediol diacrylate, 99% (reactive esters), stab. with 90ppm hydroquinone
CAS: 13048-33-4 | C12H18O4 | 226.27 g/mol
Have Questions?
Catalog NumberQuantity
ALF043203.1850 g
Catalog number ALF043203.18
Price (MYR)
357.00
EA
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
357.00
EA
Add to cart
Specifications
Chemical Name or Material1,6-Hexanediol diacrylate
Name NoteStabilized with 90ppm hydroquinone
CAS13048-33-4
Health Hazard 1H315-H317-H319
Health Hazard 2GHS H Statement
H315-H319-H317
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
View more
1,6-Hexanediol diacrylate is used as a functional monomer for polymers. It acts as a cross linking agent between the molecular chains of polymers. Further, it is used in adhesives, sealants, alkyd coatings, elastomers, photopolymers, and inks for improved adhesion, hardness, abrasion and heat resistance.