Nickel wire, 0.25mm (0.01in) dia, annealed, 99.98% (metals basis)
Nickel wire, 0.25mm (0.01in) dia, annealed, 99.98% (metals basis)
Nickel wire, 0.25mm (0.01in) dia, annealed, 99.98% (metals basis)
Nickel wire, 0.25mm (0.01in) dia, annealed, 99.98% (metals basis)
Thermo Scientific Chemicals
Nickel wire, 0.25mm (0.01in) dia, annealed, 99.98% (metals basis)
Form: Approx.0.44g/m, Temper: soft | CAS: 7440-02-0 | Ni | 58.69 g/mol
Have Questions?
Catalog NumberQuantity
ALF043131.G55 m
Catalog number ALF043131.G5
Price (MYR)
772.00
EA
Add to cart
Quantity:
5 m
Request bulk or custom format
Price (MYR)
772.00
EA
Add to cart
Specifications
Chemical Name or MaterialNickel wire, Annealed
ColorSilver to Gray
CAS7440-02-0
Health Hazard 1H317-H351-H372
Health Hazard 3P201-P202-P260-P261-P264b-P270-P272-P280-P302+P352-P308+P313-P333+P313-P363-P501c