Titanium powder, -60+100 mesh, 99.5% (metals basis)
Titanium powder, -60+100 mesh, 99.5% (metals basis)
Titanium powder, -60+100 mesh, 99.5% (metals basis)
Titanium powder, -60+100 mesh, 99.5% (metals basis)
Thermo Scientific Chemicals
Titanium powder, -60+100 mesh, 99.5% (metals basis)
CAS: 7440-32-6 | Ti | 47.87 g/mol
Have Questions?
Catalog NumberQuantity
ALF043105.22100 g
Catalog number ALF043105.22
Price (MYR)
471.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
471.00
EA
Add to cart
Specifications
Chemical Name or MaterialTitanium powder
CAS7440-32-6
Health Hazard 1H250-H500
Health Hazard 2GHS H Statement
H252
Self-heating in large quantities
may catch fire.
Health Hazard 3P210-P222-P280-P335+P334-P370+P378q