Copper foil, 0.05mm (0.002in) thick, Puratronic™, 99.9999% (metals basis)
Copper foil, 0.05mm (0.002in) thick, Puratronic™, 99.9999% (metals basis)
Copper foil, 0.05mm (0.002in) thick, Puratronic™, 99.9999% (metals basis)
Copper foil, 0.05mm (0.002in) thick, Puratronic™, 99.9999% (metals basis)
Thermo Scientific Chemicals
Copper foil, 0.05mm (0.002in) thick, Puratronic™, 99.9999% (metals basis)
Form: Approx. 1.1g/50 x 50mm | CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Catalog NumberQuantity
ALF042972.GH100 x 100 mm
Catalog number ALF042972.GH
Price (MYR)
1,633.00
EA
Add to cart
Quantity:
100 x 100 mm
Request bulk or custom format
Price (MYR)
1,633.00
EA
Add to cart
Specifications
Chemical Name or MaterialCopper
CAS7440-50-8
AppearanceCopper colored foil
Density8.94g/cm3