Disodium terephthalate, 99+%
Disodium terephthalate, 99+%
Disodium terephthalate, 99+%
Disodium terephthalate, 99+%
Thermo Scientific Chemicals
Disodium terephthalate, 99+%
CAS: 10028-70-3 | C8H4Na2O4 | 210.096 g/mol
Have Questions?
Catalog NumberQuantity
ALF042946.22100 g
Catalog number ALF042946.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialDisodium terephthalate
CAS10028-70-3
Recommended StorageAmbient temperatures
EINECS Number233-075-5
TSCANo
View more
Disodium terephthalate and its various derivatives are synthesized via simple acid-base chemistry for anode materials in Na ion batteries. They show excellent electrochemical performance, including little capacity fading over 90 cycles, ideal redox potential, and excellent rate performance, making them promising candidates for Na ion batteries.